3-Bromo-N-cyclopropyl-5-(trifluoromethoxy)benzamide ≥95%

Supplier: AOBChem USA

Warning

58264-500MG 58264-1G 58264-5G 58264-25G
77330-206EA 264.71 USD
77330-206 77354-242 77366-470 77388-196
3-Bromo-N-cyclopropyl-5-(trifluoromethoxy)benzamide ≥95%
3-Bromo-N-cyclopropyl-5-(trifluoromethoxy)benzamide

Precaución: for Research Use Only

Formula: C₁₁H₉BrF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD32633133
CAS Number: 2752240-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR