3-Bromo-N,N-dimethyl-5-(trifluoromethoxy)benzamide ≥97%

Supplier: AOBChem USA

Warning

58263-500MG 58263-1G 58263-5G 58263-25G
77330-204EA 264.71 USD
77330-204 77354-240 77366-468 77388-194
3-Bromo-N,N-dimethyl-5-(trifluoromethoxy)benzamide ≥97%
3-Bromo-N,N-dimethyl-5-(trifluoromethoxy)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD32633139
CAS Number: 2751326-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR