3-Bromo-N-methyl-5-(trifluoromethoxy)benzamide ≥97%

Supplier: AOBChem USA

Warning

58262-500MG 58262-1G 58262-5G 58262-25G
77330-202EA 264.71 USD
77330-202 77354-238 77366-466 77388-192
3-Bromo-N-methyl-5-(trifluoromethoxy)benzamide ≥97%
3-Bromo-N-methyl-5-(trifluoromethoxy)benzamide

Precaución: for Research Use Only

Formula: C₉H₇BrF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD32633130
CAS Number: 2752028-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR