3',5'-Dichloro-4'-isopropoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58254-500MG 58254-1G 58254-5G
77338-380EA 484.1 USD
77338-380 77354-236 77378-392
3',5'-Dichloro-4'-isopropoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3',5'-Dichloro-4'-isopropoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₄Cl₂O₂
MW: 309.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758554
CAS Number: 2764730-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR