3,5-Dichloro-4'-fluoro-4-isopropoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58253-500MG 58253-1G 58253-5G
77335-602EA 360.5 USD
77335-602 77354-234 77378-390
3,5-Dichloro-4'-fluoro-4-isopropoxy-1,1'-biphenyl ≥97%
3,5-Dichloro-4'-fluoro-4-isopropoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₃Cl₂FO
MW: 299.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758553
CAS Number: 2807464-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR