3',5'-Dichloro-4'-ethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58243-500MG 58243-1G 58243-5G
77336-828EA 394.49 USD
77336-828 77354-232 77378-388
3',5'-Dichloro-4'-ethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3',5'-Dichloro-4'-ethoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₂Cl₂O₂
MW: 295.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758544
CAS Number: 2764730-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR