3,5-Dichloro-4-ethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58239-500MG 58239-1G 58239-5G
77336-102EA 375.95 USD
77336-102 77354-226 77378-382
3,5-Dichloro-4-ethoxy-1,1'-biphenyl ≥97%
3,5-Dichloro-4-ethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD34758541
CAS Number: 2764728-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR