2-(Benzyloxy)-1,3-dichloro-5-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

58238-500MG 58238-1G 58238-5G 58238-25G
77330-198EA 160.68 USD
77330-198 77345-800 77366-464 77388-190
2-(Benzyloxy)-1,3-dichloro-5-iodobenzene ≥95%
2-(Benzyloxy)-1,3-dichloro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉Cl₂IO
MW: 379.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564544
CAS Number: 1681028-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR