3,5-Dichloro-4'-fluoro-4-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58234-500MG 58234-1G 58234-5G 58234-25G
77330-194EA 225.57 USD
77330-194 77354-224 77366-460 77388-186
3,5-Dichloro-4'-fluoro-4-methoxy-1,1'-biphenyl ≥97%
3,5-Dichloro-4'-fluoro-4-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD34758538
CAS Number: 1841-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR