3,5-Dichloro-4-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58231-500MG 58231-1G 58231-5G 58231-25G
77330-190EA 225.57 USD
77330-190 77354-220 77366-458 77388-182
3,5-Dichloro-4-methoxy-1,1'-biphenyl ≥97%
3,5-Dichloro-4-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀Cl₂O
MW: 253.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758536
CAS Number: 74298-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR