3-Bromo-5-(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58226-500MG 58226-1G 58226-25G 58226-5G
77330-188EA 300.76 USD
77330-188 77354-216 77388-180 77378-376
3-Bromo-5-(trifluoromethoxy)-1,1'-biphenyl ≥97%
3-Bromo-5-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD34758532
CAS Number: 2755722-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR