(3-Iodo-5-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

58224-500MG 58224-1G 58224-5G 58224-25G
77330-186EA 194.67 USD
77330-186 77354-214 77366-456 77388-178
(3-Iodo-5-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%
(3-Iodo-5-(trifluoromethoxy)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₆F₃IOS
MW: 334.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758531
CAS Number: 2385946-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR