3-(Methylthio)-5-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58222-500MG 58222-1G 58222-5G 58222-25G
77330-184EA 270.89 USD
77330-184 77354-212 77366-454 77388-176
3-(Methylthio)-5-(trifluoromethoxy)benzaldehyde ≥97%
3-(Methylthio)-5-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂S
MW: 236.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758529
CAS Number: 2167962-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR