3-(Methylthio)-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

58219-500MG 58219-1G 58219-25G 58219-5G
77330-182EA 284.28 USD
77330-182 77354-210 77388-174 77378-374
3-(Methylthio)-5-(trifluoromethoxy)benzoic acid ≥97%
3-(Methylthio)-5-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD32701836
CAS Number: 2749691-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR