(3-Iodo-5-(trifluoromethoxy)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

58211-500MG 58211-1G 58211-5G 58211-25G
77330-180EA 259.56 USD
77330-180 77354-208 77366-452 77388-172
(3-Iodo-5-(trifluoromethoxy)phenyl)methanol ≥97%
(3-Iodo-5-(trifluoromethoxy)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₆F₃IO₂
MW: 318.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758521
CAS Number: 2169423-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR