3-(2-Fluorophenyl)-3-hydroxypropanoic acid ≥95%

Supplier: AOBChem USA

Warning

58195-500MG 58195-1G 58195-5G
77339-002EA 543.84 USD
77339-002 77354-194 77378-366
3-(2-Fluorophenyl)-3-hydroxypropanoic acid ≥95%
3-(2-Fluorophenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD16302335
CAS Number: 60187-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR