3-(4-Chlorophenyl)-3-hydroxypropanoic acid ≥95%

Supplier: AOBChem USA

Warning

58194-500MG 58194-1G 58194-5G 58194-25G
77330-170EA 215.27 USD
77330-170 77354-192 77366-444 77388-162
3-(4-Chlorophenyl)-3-hydroxypropanoic acid ≥95%
3-(4-Chlorophenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD16304917
CAS Number: 25209-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR