(4-Ethoxy-3-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

58181-500MG 58181-1G 58181-5G 58181-25G
77330-166EA 232.78 USD
77330-166 77354-190 77366-438 77388-160
(4-Ethoxy-3-fluorophenyl)methanol ≥97%
(4-Ethoxy-3-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11190336
CAS Number: 1039954-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR