3'-Bromo-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58167-500MG 58167-1G 58167-5G
77336-140EA 378.01 USD
77336-140 77354-180 77378-354
3'-Bromo-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3'-Bromo-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₈BrF₃O
MW: 329.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758498
CAS Number: 2755723-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR