3-Bromo-4'-fluoro-5-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58166-500MG 58166-1G 58166-5G
77338-378EA 484.1 USD
77338-378 77354-178 77378-352
3-Bromo-4'-fluoro-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
3-Bromo-4'-fluoro-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₄
MW: 319.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758497
CAS Number: 2755719-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR