1,2-Dibromo-4-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

58159-500MG 58159-1G 58159-5G 58159-25G
77330-156EA 158.62 USD
77330-156 77345-740 77366-434 77388-156
1,2-Dibromo-4-methoxy-3-methylbenzene ≥97%
1,2-Dibromo-4-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
MW: 279.957 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758492
CAS Number: 2385829-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR