3,4-Dibromo-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

58158-500MG 58158-1G 58158-5G 58158-25G
77330-154EA 118.45 USD
77330-154 77343-564 77366-432 77378-346
3,4-Dibromo-2-methylphenol ≥97%
3,4-Dibromo-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
MW: 265.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758491
CAS Number: 55289-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR