3,6-Dibromo-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

58156-500MG 58156-1G 58156-5G 58156-25G
77330-152EA 99.91 USD
77330-152 77342-840 77366-430 77378-344
3,6-Dibromo-2-methylphenol ≥97%
3,6-Dibromo-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
Storage Temperature: Ambient
MDL Number: MFCD18969994
CAS Number: 52045-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR