1,3-Dibromo-5-isobutyl-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

58125-500MG 58125-1G 58125-5G 58125-25G
77330-148EA 258.53 USD
77330-148 77354-166 77366-426 77388-152
1,3-Dibromo-5-isobutyl-2-methoxybenzene ≥97%
1,3-Dibromo-5-isobutyl-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₄Br₂O
Storage Temperature: Ambient
MDL Number: MFCD34564535
CAS Number: 2734776-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR