1,3-Dibromo-2-ethoxy-5-isobutylbenzene ≥95%

Supplier: AOBChem USA

Warning

58124-500MG 58124-1G 58124-25G 58124-5G
77330-146EA 287.37 USD
77330-146 77354-164 77388-150 77378-338
1,3-Dibromo-2-ethoxy-5-isobutylbenzene ≥95%
1,3-Dibromo-2-ethoxy-5-isobutylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₆Br₂O
MW: 336.063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564534
CAS Number: 2734778-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR