Methyl 5-isobutyl-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

58119-500MG 58119-1G 58119-5G
77336-888EA 396.55 USD
77336-888 77354-160 77378-336
Methyl 5-isobutyl-2-methoxybenzoate ≥97%
Methyl 5-isobutyl-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30830869
CAS Number: 1807918-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR