5-Isobutyl-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

58118-500MG 58118-1G 58118-5G
77337-626EA 431.57 USD
77337-626 77354-158 77378-334
5-Isobutyl-2-methoxybenzoic acid ≥97%
5-Isobutyl-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24487719
CAS Number: 343316-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR