2-Iodo-4-isobutyl-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

58115-500MG 58115-1G 58115-5G
77337-722EA 438.78 USD
77337-722 77354-156 77378-332
2-Iodo-4-isobutyl-1-methoxybenzene ≥97%
2-Iodo-4-isobutyl-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅IO
MW: 290.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758461
CAS Number: 2764728-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR