2-Bromo-4-isobutyl-1-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

58110-500MG 58110-1G 58110-5G
77330-140EA 348.14 USD
77330-140 77354-152 77378-328
2-Bromo-4-isobutyl-1-(methoxymethoxy)benzene ≥97%
2-Bromo-4-isobutyl-1-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrO₂
MW: 273.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758456
CAS Number: 2755721-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR