2-Fluoro-N,5-dimethoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

58109-500MG 58109-1G 58109-5G 58109-25G
77330-138EA 153.47 USD
77330-138 77345-512 77366-422 77388-144
2-Fluoro-N,5-dimethoxybenzamide ≥97%
2-Fluoro-N,5-dimethoxybenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₃
MW: 199.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564532
CAS Number: 2734778-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR