(4-Methyl-[1,1'-biphenyl]-2-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

58099-500MG 58099-1G 58099-25G 58099-5G
77330-128EA 297.67 USD
77330-128 77354-144 77388-134 77378-326
(4-Methyl-[1,1'-biphenyl]-2-yl)boronic acid ≥97%
(4-Methyl-[1,1'-biphenyl]-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BO₂
MW: 212.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32219532
CAS Number: 2243975-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR