(5-Chloro-2-ethoxy-3-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

58098-500MG 58098-1G 58098-5G
77339-390EA 584.01 USD
77339-390 77354-142 77378-324
(5-Chloro-2-ethoxy-3-fluorophenyl)(methyl)sulfane ≥97%
(5-Chloro-2-ethoxy-3-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀ClFOS
Storage Temperature: Ambient
MDL Number: MFCD34758455
CAS Number: 2710093-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR