(2-Fluoro-6-iodo-3-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

58094-500MG 58094-1G 58094-5G
77330-126EA 334.75 USD
77330-126 77354-140 77378-322
(2-Fluoro-6-iodo-3-methylphenyl)methanol ≥97%
(2-Fluoro-6-iodo-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈FIO
MW: 266.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29040754
CAS Number: 1936043-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR