(3-Chloro-4-fluoro-5-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

58093-500MG 58093-1G 58093-5G 58093-25G
77330-124EA 192.61 USD
77330-124 77354-138 77366-412 77388-132
(3-Chloro-4-fluoro-5-iodophenyl)methanol ≥97%
(3-Chloro-4-fluoro-5-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34551662
CAS Number: 2385117-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR