3-Chloro-4-fluoro-5-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58092-500MG 58092-1G 58092-5G 58092-25G
77330-122EA 150.38 USD
77330-122 77345-362 77366-410 77388-130
3-Chloro-4-fluoro-5-iodobenzaldehyde ≥97%
3-Chloro-4-fluoro-5-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFIO
Storage Temperature: Ambient
MDL Number: MFCD34758451
CAS Number: 2387135-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR