(3-Chloro-4-fluoro-5-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

58083-500MG 58083-1G 58083-5G
77338-874EA 527.36 USD
77338-874 77354-136 77378-318
(3-Chloro-4-fluoro-5-methylphenyl)methanol ≥95%
(3-Chloro-4-fluoro-5-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO
MW: 174.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28519763
CAS Number: 1785492-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR