Ethyl 3-chloro-4-fluoro-5-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

58081-500MG 58081-1G 58081-5G
77336-886EA 396.55 USD
77336-886 77354-134 77378-316
Ethyl 3-chloro-4-fluoro-5-methylbenzoate ≥95%
Ethyl 3-chloro-4-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD24108593
CAS Number: 1536923-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR