Methyl 3-chloro-4-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

58080-500MG 58080-1G 58080-5G
77336-826EA 394.49 USD
77336-826 77354-132 77378-314
Methyl 3-chloro-4-fluoro-5-methylbenzoate ≥97%
Methyl 3-chloro-4-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24108592
CAS Number: 1509153-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR