3-Chloro-4-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

58079-500MG 58079-1G 58079-5G
77337-720EA 438.78 USD
77337-720 77354-130 77378-312
3-Chloro-4-fluoro-5-methylbenzoic acid ≥97%
3-Chloro-4-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712115
CAS Number: 1427392-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR