3-Chloro-4-fluoro-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

58071-500MG 58071-1G 58071-5G 58071-25G
77330-114EA 258.53 USD
77330-114 77354-128 77366-402 77388-124
3-Chloro-4-fluoro-5-(methylthio)benzoic acid ≥97%
3-Chloro-4-fluoro-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂S
Storage Temperature: Ambient
MDL Number: MFCD34758442
CAS Number: 2755717-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR