3-Chloro-4-fluoro-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58069-500MG 58069-1G 58069-5G 58069-25G
77330-112EA 257.5 USD
77330-112 77354-126 77366-400 77388-122
3-Chloro-4-fluoro-5-(methylthio)benzaldehyde ≥97%
3-Chloro-4-fluoro-5-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFOS
MW: 204.649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758440
CAS Number: 2755721-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR