Methyl 2,4-dimethyl-5-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

58062-500MG 58062-1G 58062-5G 58062-25G
77330-100EA 220.42 USD
77330-100 77354-118 77366-390 77388-110
Methyl 2,4-dimethyl-5-(methylthio)benzoate ≥97%
Methyl 2,4-dimethyl-5-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂S
MW: 210.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758434
CAS Number: 2755721-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR