Methyl 6-(4-formylphenyl)nicotinate ≥95%

Supplier: AOBChem USA

Warning

58051-500MG 58051-1G 58051-5G 58051-25G
77330-094EA 257.5 USD
77330-094 77354-110 77366-386 77388-106
Methyl 6-(4-formylphenyl)nicotinate ≥95%
Methyl 6-(4-formylphenyl)nicotinate

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₃
MW: 241.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758426
CAS Number: 2755724-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR