Methyl 6-(4-fluorophenyl)nicotinate ≥97%

Supplier: AOBChem USA

Warning

58047-500MG 58047-1G 58047-5G 58047-25G
77330-092EA 207.03 USD
77330-092 77354-108 77366-384 77388-104
Methyl 6-(4-fluorophenyl)nicotinate ≥97%
Methyl 6-(4-fluorophenyl)nicotinate

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO₂
MW: 231.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836551
CAS Number: 149500-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR