Methyl 6-(3-fluorophenyl)nicotinate ≥97%

Supplier: AOBChem USA

Warning

58044-500MG 58044-1G 58044-5G
77330-090EA 311.06 USD
77330-090 77354-106 77378-304
Methyl 6-(3-fluorophenyl)nicotinate ≥97%
Methyl 6-(3-fluorophenyl)nicotinate

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO₂
MW: 231.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27578640
CAS Number: 1820609-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR