4-(5-(Hydroxymethyl)pyridin-2-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

58043-500MG 58043-1G 58043-5G 58043-25G
77330-088EA 187.46 USD
77330-088 77354-104 77366-382 77388-102
4-(5-(Hydroxymethyl)pyridin-2-yl)phenol ≥97%
4-(5-(Hydroxymethyl)pyridin-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂
MW: 201.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564526
CAS Number: 2734776-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR