6-(4-Methylphenyl)nicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-(p-Tolyl)nicotinaldehyde

58041-500MG 58041-1G 58041-5G 58041-25G
77330-086EA 258.53 USD
77330-086 77354-102 77366-380 77388-100
6-(4-Methylphenyl)nicotinaldehyde ≥97%
6-(4-Methylphenyl)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115442
CAS Number: 898795-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR