1-(2-(Benzyloxy)-4-methoxyphenyl)-2,2-dichloroethanol ≥95%

Supplier: AOBChem USA

Warning

58040-500MG 58040-1G 58040-5G 58040-25G
77330-084EA 204.97 USD
77330-084 77354-100 77366-378 77388-098
1-(2-(Benzyloxy)-4-methoxyphenyl)-2,2-dichloroethanol ≥95%
1-(2-(Benzyloxy)-4-methoxyphenyl)-2,2-dichloroethanol

Precaución: for Research Use Only

Formula: C₁₆H₁₆Cl₂O₃
MW: 327.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564525
CAS Number: 2734777-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR