4-(5-Isopropoxypyridin-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58014-500MG 58014-1G 58014-5G
77338-344EA 479.98 USD
77338-344 77354-098 77378-302
4-(5-Isopropoxypyridin-3-yl)benzaldehyde ≥97%
4-(5-Isopropoxypyridin-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₂
MW: 241.285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25743927
CAS Number: 1545014-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR