(5-Bromo-2,3-dimethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

58008-500MG 58008-1G 58008-5G
77335-992EA 372.86 USD
77335-992 77354-090 77378-294
(5-Bromo-2,3-dimethoxyphenyl)methanol ≥97%
(5-Bromo-2,3-dimethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₃
MW: 247.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04210208
CAS Number: 86232-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR